Invitrogen
Manufacturer Code:MA527985
Catalog # PIMA527985
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | Chk1S317-F10 |
Immunogen | A synthetic phospho-peptide corresponding to residues surrounding Ser317 of human phospho Chk1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: Cell cycle checkpoint kinase; checkpoint kinase 1 homolog; Checkpoint kinase-1; Checkpoint, S. pombe, homolog of, 1; CHK1 checkpoint homolog; Chk1-S; rad27 homolog; Serine/threonine-protein kinase Chk1
Gene Aliases: C85740; CHEK1; CHK1; rad27
UniProt ID: (Human) O14757, (Mouse) O35280
Entrez Gene ID: (Human) 1111, (Mouse) 12649
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.