Invitrogen
Manufacturer Code:PA517873
Catalog # PIPA517873
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic phosphopeptide corresponding to residues surrounding pTyr15 of human cdc2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CDC2; cdc2 kinase; CDK1; CDK1-A; Cdk1-PA; cell cycle controller CDC2; cell cycle p34 CDC2 kinase protein; cell division control; Cell division control protein 2 homolog; Cell division control protein 2 homolog 1; Cell division control protein 2-A; cell division cycle 2 homolog A; cell division cycle 2 like; cell division cycle 2, G1 to S and G2 to M; Cell division cycle control protein 2; cell division cycle control protein 2a; Cell division protein kinase 1; Cell division protein kinase 1-A; CG5363-PA; cyclin dependent kinase; cyclin-dependent kinase; Cyclin-dependent kinase 1; Cyclin-dependent kinase 1-A; LD38718p; maturation-promoting factor; OTTHUMP00000019660 cell cycle controller CDC2 cell division control protein 2 homolog cell division cycle 2 G1 to S and G2 to M cell division protein kinase 1 p34 protein kinase; p34 protein kinase; p34 protein kinase 1; protein 2
Gene Aliases: cdc; 5363; cdc-2; CDC2; cdc2-a; CDC28A; Cdc2a; cdc2Dm; cdc2x1.1; CDCDm; Cdk-1; CDK1; cdk1-a; cdk1-b; CDK1/CDC2; CDKN1; CG5363; Dcdc2; dCdk1; Dm cdc2; DmCdc2; DmCdk1; Dmel\CG5363; Dmel_CG5363; group 4; l(2)31Eh; MPF; p34<CDC2>; P34CDC2; PSTAIR; xcdc2; XELAEV_18036802mg
UniProt ID: (Human) P06493, (Rat) P39951, (Fruit fly) P23572, (Mouse) P11440, (Xenopus) P35567
Entrez Gene ID: (Human) 983, (Rat) 54237, (Fruit fly) 34411, (Mouse) 12534, (Xenopus) 380246
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.