Invitrogen
Manufacturer Code:PA599351
Catalog # PIPA599351
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Integrin beta3 around the phosphorylation site of Tyr785. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: antigen CD61; antigen CD61 BDPLT16 BDPLT2 beta3 integrin CD61 EGK_08849 glycoprotein GPIIIa glycoprotein IIIa GP3A GPIIIa GT INGRB3 integrin beta 3 integrin beta 3 (Cd61) integrin beta-3 integrin beta-3 subunit integrin subunit beta 3 integrin beta 3 integrin beta 3 (platelet glycoprotein IIIa antigen CD61) ITGB3 platelet glycoprotein IIIa integrin subunit beta 3 platelet gpIIIa platelet membrane glycoprotein IIIa; CD61; GPIIIa; integrin beta 3; integrin beta 3 (Cd61); Integrin beta-3; integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61); platelet gpIIIa; Platelet membrane glycoprotein IIIa
Gene Aliases: BDPLT16; BDPLT2; CD61; GP3A; GPIIIa; GT; INGRB3; ITGB3
UniProt ID: (Human) P05106, (Rat) Q8R2H2, (Mouse) O54890
Entrez Gene ID: (Human) 3690, (Rat) 29302, (Mouse) 16416
Molecular Function:
cell adhesion molecule
extracellular matrix glycoprotein
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.