Invitrogen
Manufacturer Code:PA538458
Catalog # PIPA538458
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic phosphopeptide derived from human Coagulation Factor III around the phosphorylation site of Ser290 (E-N-SP-P-L) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD142; Coagulation factor III; coagulation factor III (thromboplastin, tissue factor); F3 Coagulation factor 3 TF TFA Coagulation Factor III; TF; Thromboplastin; Tissue factor
Gene Aliases: AA409063; CD142; Cf-3; Cf3; F3; TF; TFA
UniProt ID: (Human) P13726, (Mouse) P20352
Entrez Gene ID: (Human) 2152, (Mouse) 14066
Molecular Function:
cytokine receptor
defense/immunity protein
receptor
type I cytokine receptor
type II cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.