Invitrogen
Manufacturer Code:PA5105011
Catalog # PIPA5105011
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CaMK4 around the phosphorylation site of Thr200. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A430110E23Rik AI666733 brain Ca(2+)-calmodulin-dependent protein kinase type IV brain Ca++-calmodulin-dependent protein kinase type IV Ca2+/calmodulin-dependent protein kinase type IV/Gr Calcium/calmoduli calcium/calmodulin dependent protein kinase IV calcium/calmodulin-dependent protein kinase IV calcium/calmodulin-dependent protein kinase IV L homeolog calcium/calmodulin-dependent protein kinase type IV calcium/calmodulin-dependent protein kinase type IV catalytic chain Calmodulin-dependent protein kinase IV calspermin CAM kinase- GR CAM kinase IV CAM kinase-GR caMK CaMK IV camk4 camk4.L camk4-A CAMK-GR CaMKIV CaMKIV/Gr Ccdpk D18Bwg0362e EC 2.7.11.17 KCC4 kinase CaMK4 OTTHUMP00000222818 RATCCDPK xCaM XELAEV_18008011mg; brain Ca(2+)-calmodulin-dependent protein kinase type IV; brain Ca++-calmodulin-dependent protein kinase type IV; Ca2+/calmodulin-dependent protein kinase type IV/Gr; calcium/calmodulin-dependent protein kinase IV; Calcium/calmodulin-dependent protein kinase type IV; calcium/calmodulin-dependent protein kinase type IV catalytic chain; Calmodulin-dependent protein kinase IV; Calspermin; CAM kinase IV; CAM kinase- GR; CAM kinase-GR; CaMK IV
Gene Aliases: A430110E23Rik; AI666733; CAMK; CaMK IV; CAMK-GR; CAMK4; CAMKIV; CaMKIV/Gr; Ccdpk; D18Bwg0362e; RATCCDPK
UniProt ID: (Human) Q16566, (Rat) P13234
Entrez Gene ID: (Human) 814, (Mouse) 12326, (Rat) 25050
Molecular Function:
cytoskeletal protein
kinase
microtubule family cytoskeletal protein
non-motor microtubule binding protein
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.