Invitrogen
Manufacturer Code:PA5105158
Catalog # PIPA5105158
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CAD around the phosphorylation site of Ser1859. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2410008J01Rik 38E.19 anon-WO2004063362.83 Aspartate carbamoyltransferase AU018859 cad CAD protein CAD trifunctional protein cad-PA cad-PB cad-PC carbamoyl-phosphate synthetase 2 aspartate transcarbamylase and dihydroorotase carbamyl phosphatate synthetase 2 caudal cd CDG1Z CG1759 CG1759 gene product from transcript CG1759-RC CG1759-PA CG1759-PB CG1759-PC Cpad Dihydroorotase DmelCG1759 Dmel_CG1759 Glutamine-dependent carbamoyl-phosphate synthase Homeotic protein caudal multifunctional protein CAD S67; Aspartate carbamoyltransferase; CAD protein; CAD trifunctional protein; carbamyl phosphatate synthetase 2; Dihydroorotase; Glutamine-dependent carbamoyl-phosphate synthase; multifunctional protein CAD
Gene Aliases: 2410008J01Rik; AU018859; CAD; CDG1Z; Cpad
UniProt ID: (Human) P27708, (Mouse) B2RQC6
Entrez Gene ID: (Human) 790, (Mouse) 69719, (Rat) 24240
Molecular Function:
ligase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.