Invitrogen
Manufacturer Code:PA5117261
Catalog # PIPA5117261
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A phospho synthetic peptide corresponding to residues surrounding S291 of Mouse CGAS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2'3'-cGAMP synthase; 2'3'-cGAMP synthase C6orf150 cf150 cGAMP synthase CGAS Cyclic GMP-AMP synthase E330016A19Rik h-cGAS Mab-21 domain containing 1 mab-21 domain-containing protein 1 Mb21d1 protein MB21D1; cGAMP synthase; Cyclic GMP-AMP synthase; Mab-21 domain-containing protein 1; protein MB21D1
Gene Aliases: C6orf150; CGAS; E330016A19Rik; h-cGAS; MB21D1
UniProt ID: (Human) Q8N884, (Mouse) Q8C6L5
Entrez Gene ID: (Human) 115004, (Mouse) 214763
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.