Invitrogen
Manufacturer Code:PA517708
Catalog # PIPA517708
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic phosphopeptide corresponding to residues surrounding pTyr177 of human Bcr |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BCR/FGFR1 chimera protein; breakpoint cluster region; breakpoint cluster region homolog; Breakpoint cluster region protein; breakpoint cluster region protein renal carcinoma antigen NY-REN-26; FGFR1/BCR chimera protein; Renal carcinoma antigen NY-REN-26
Gene Aliases: 5133400C09Rik; AI561783; AI853148; ALL; BCR; BCR1; CML; D22S11; D22S662; Kiaa3017; mKIAA3017; PHL
UniProt ID: (Human) P11274, (Mouse) Q6PAJ1
Entrez Gene ID: (Human) 613, (Mouse) 110279
Molecular Function: G-protein modulator enzyme modulator guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.