Invitrogen
Manufacturer Code:PA5104784
Catalog # PIPA5104784
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human BCL-2 around the phosphorylation site of Thr69. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Apoptosis regulator Bcl-2; Apoptosis regulator Bcl-2 AW986256 B cell leukemia/lymphoma 2 B cell lymphoma 2 associated oncogene B-cell CLL/lymphoma 2 B-cell leukemia/lymphoma 2 Bcl 2 BCL2 bcl-2 BCL2 apoptosis regulator BCL2 apoptosis regulator Bcl2-like protein C430015F12Rik D630044D05Rik D830018M01Rik OTTHUMP00000163680 PPP1R50 protein phosphatase 1 regulatory subunit 50; B cell lymphoma 2 associated oncogene; B-cell CLL/lymphoma 2; B-cell leukemia/lymphoma 2; Bcl2-like protein; protein phosphatase 1, regulatory subunit 50
Gene Aliases: AW986256; Bcl-2; BCL2; C430015F12Rik; D630044D05Rik; D830018M01Rik; PPP1R50
UniProt ID: (Human) P10415, (Rat) P49950, (Mouse) P10417
Entrez Gene ID: (Human) 596, (Rat) 24224, (Mouse) 12043
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.