Invitrogen
Manufacturer Code:PA538732
Catalog # PIPA538732
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic phosphopeptide derived from human AurB around the phosphorylation site of Tyr12 (W-P-YP-G-R) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AIM-1; ARK-2; Aurora 1; aurora B; Aurora kinase B; Aurora kinase B Aurora 1 Aurora- and IPL1-like midbody-associated protein 1 AIM-1 Aurora/IPL1-related kinase 2 ARK-2 Aurora-related kinase 2 STK-1 Serine/threonine-protein kinase 12 Serine/threonine-protein kinase 5Serine/threonine-protein kinase aurora-B AURKB AIK2 AIM1 AIRK2 ARK2 STK1 STK12 STK5 Aurora kinase C Aurora 3 Aurora/IPL1-related kinase 3 ARK-3 Aurora-related kinase 3 Aurora/IPL1/Eg2 protein 2 Serine/threonine-protein kinase 13 Serine/threonine-protein kinase aurora-C AURKC AIE2 AIK3 AIRK3 ARK3 STK13; aurora kinase B-Sv1; aurora kinase B-Sv2; Aurora- and IPL1-like midbody-associated protein 1; aurora-1; aurora-B; aurora-related kinase 2; Aurora/IPL1-related kinase 2; protein phosphatase 1, regulatory subunit 48; serine/threonine kinase 12; serine/threonine kinase 5; Serine/threonine-protein kinase 12; Serine/threonine-protein kinase 5; Serine/threonine-protein kinase aurora-B; STK-1
Gene Aliases: AIK2; AIM-1; AIM1; AIRK2; AL022959; ARK-2; ARK2; AurB; AURKB; aurkb-sv1; aurkb-sv2; IPL1; PPP1R48; STK-1; STK1; STK12; STK5
UniProt ID: (Human) C7G533, (Mouse) O70126, (Rat) O55099
Entrez Gene ID: (Human) 9212, (Mouse) 20877, (Rat) 114592
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.