Invitrogen
Manufacturer Code:MA514904
Catalog # PIMA514904
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | F.131.2 |
Immunogen | Synthetic phosphopeptide corresponding to residues around pThr288 of human Aurora A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: ARK-1; ARK-1 IPL1-related kinase OTTHUMP00000031341 OTTHUMP00000166072 aurora-related kinase 1 aurora/IPL1-like kinase aurora/IPL1-related kinase 1 breast tumor-amplified kinase breast-tumor-amplified kinase hARK1 serine/threonine kinase 6 serine/threonine protein kinase 15 serine/threonine-protein kinase 15 serine/threonine-protein kinase 6 serine/threonine-protein kinase aurora-A; Aurora 2; Aurora kinase A; aurora/IPL1-like kinase; Aurora/IPL1-related kinase 1; Breast tumor-amplified kinase; protein phosphatase 1, regulatory subunit 47; serine/threonine protein kinase 15; Serine/threonine-protein kinase 15; Serine/threonine-protein kinase 6; Serine/threonine-protein kinase aurora-A
Gene Aliases: AIK; AIRK1; ARK1; AURA; AURKA; AYK1; BTAK; IAK1; PPP1R47; STK15; STK6; STK7
UniProt ID: (Human) O14965
Entrez Gene ID: (Human) 6790
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.