Invitrogen
Manufacturer Code:PA5105959
Catalog # PIPA5105959
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Androgen Receptor around the phosphorylation site of Tyr535. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AIS Andr androgen nuclear receptor variant 2 androgen receptor androgen receptor (Testicular feminization) same as Tfm androgen receptor splice variant 4b AR AR8 AR-A AW320017 DHTR Dihydrotestosterone receptor HUMARA HYSP1 KD NR3C4 Nuclear receptor subfamily 3 group C member 4 RP11-383C12.1 SBMA SMAX1 TFM Variant AR45; androgen nuclear receptor variant 2; Androgen receptor; androgen receptor (Testicular feminization), same as Tfm; androgen receptor splice variant 4b; Dihydrotestosterone receptor; Nuclear receptor subfamily 3 group C member 4
Gene Aliases: AIS; Andr; AR; AR8; AW320017; DHTR; HUMARA; HYSP1; KD; NR3C4; SBMA; SMAX1; TFM
UniProt ID: (Human) P10275, (Rat) P15207, (Mouse) P19091
Entrez Gene ID: (Human) 367, (Rat) 24208, (Mouse) 11835
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.