Invitrogen
Manufacturer Code:PA5106161
Catalog # PIPA5106161
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Sodium Potassium ATPase alpha 1 around the phosphorylation site of Tyr260. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: (Na+ + K+)-ATPase (Na+ K+)-ATPase alpha-subunit Atp1a1 atp1a1-a atp1a1a.4 atp1a1-b ATP1B Atpa-1 ATPase Na+/K+ transporting subunit alpha 1 ATPase Na+/K+ transporting alpha 1 polypeptide ATPase Na+K+ transporting alpha 1 BC010319 K-ATPase MGC1798 Na(+)/K(+) ATPase alpha-1 subunit Na K-ATPase alpha-A catalytic polypeptide NaK-ATPase alpha-1 subunit NaK-ATPase catalytic subunit alpha-A protein Na/K ATPase alpha 1 isoform Na/K ATPase alpha 1 subunit Na+ K+ -ATPase alpha subunit Na+ /K+ ATPase alpha 1 subunit Na+ K+-ATPase beta-subunit precursor Na+/K+ ATPase Na+/K+ ATPase 1 Na+/K+ ATPase Alpha Na+/K+ -ATPase alpha 1 subunit Na+/K+ transporting alpha 1 polypeptide Na+K+ ATPase Na+K+ATPase Nkaa1b RP4-655J12.1 Sodium Potassium ATPase sodium pump 1 sodium pump subunit alpha-1 sodium/potassium-ATPase alpha-subunit 1 sodium/potassium-transporting ATPase alpha-1 chain Sodium/potassium-transporting ATPase subunit alpha-1 sodium-potassium ATPase catalytic subunit alpha-1 sodium-potassium-ATPase alpha 1 polypeptide unnamed protein product; ATPase, Na+/K+ transporting, alpha 1 polypeptide; ATPase, Na+K+ transporting, alpha 1; Na(+)/K(+) ATPase alpha-1 subunit; Na+/K+ ATPase 1; Na, K-ATPase, alpha-A catalytic polypeptide; Na,K-ATPase alpha-1 subunit; Na,K-ATPase catalytic subunit alpha-A protein; Sodium pump subunit alpha-1; sodium-potassium ATPase catalytic subunit alpha-1; sodium-potassium-ATPase, alpha 1 polypeptide; Sodium/potassium-transporting ATPase subunit alpha-1
Gene Aliases: ATP1A1; Atpa-1; BC010319; Nkaa1b
UniProt ID: (Human) P05023, (Rat) Q64609, (Mouse) Q8VDN2
Entrez Gene ID: (Human) 476, (Rat) 24211, (Mouse) 11928
Molecular Function: cation transporter hydrolase ion channel transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.