Invitrogen
Manufacturer Code:PA577871
Catalog # PIPA577871
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human ATM (phospho Ser1981). The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A-T mutated; AT mutated; Ataxia telangiectasia mutated; phospho S1981 ATM phospho ATM ATM phospho S1981 ATM serine/threonine kinase phospho ATM (S1981) ATM phospho Ser1981 ATE ATD ATC ATM phospho ATM (pSer1981) TELO1 phospho ATM (pS1981) ATM (pS1981) ATA AT1 TEL1 ATM (phospho S1981) phospho Ser1981 ATM ATDC phospho ATM (pSer1981) ATM (phospho Ser1981); Serine-protein kinase ATM; TEL1, telomere maintenance 1, homolog
Gene Aliases: AT1; ATA; ATC; ATD; ATDC; ATE; ATM; TEL1; TELO1
UniProt ID: (Human) Q13315
Entrez Gene ID: (Human) 472
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
nucleic acid binding
nucleotide kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.