Invitrogen
Manufacturer Code:PA5110151
Catalog # PIPA5110151
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A phospho specific peptide corresponding to residues surrounding T172 of human AMPK alpha. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2310008I11Rik 5&prime-AMP-activated protein kinase catalytic subunit alpha-1 5&prime-AMP-activated protein kinase catalytic subunit alpha-2 A830082D05 AAPK2 ACACA kinase Acetyl-CoA carboxylase kinase AI194361 AI450832 AL024255 AMPK AMPK subunit alpha-1 AMPK subunit alpha-2 AMPK1 AMPK2 AMPKa1 Ampka2 AMPKalpha1 AMPK-alpha-1 chain AMPKalpha2 BOS_19826 BOS_3574 C130083N04Rik HMGCR kinase Hydroxymethylglutaryl-CoA reductase kinase pAMPK PRKAA Prkaa1 PRKAA2 Tau-protein kinase PRKAA1; 5'-AMP-activated protein kinase catalytic subunit alpha-1; 5'-AMP-activated protein kinase catalytic subunit alpha-2; 5'-AMP-activated protein kinase, catalytic alpha-1 chain; 5'-AMP-activated protein kinase, catalytic alpha-2 chain; ACACA kinase; Acetyl-CoA carboxylase kinase; AMP -activate kinase alpha 1 subunit; AMP-activated protein kinase; AMP-activated protein kinase alpha-2 subunit variant 2; AMP-activated protein kinase alpha-2 subunit variant 3; AMP-activated protein kinase alpha-2 variant B; AMP-activated protein kinase, alpha 1 catalytic subunit; AMP-activated protein kinase, catalytic, alpha-1; AMPK alpha 1; AMPK alpha 2; AMPK alpha-1 chain; AMPK alpha-2 chain; AMPK subunit alpha-1; AMPK subunit alpha-2; AMPK-alpha-2 chain; HMGCR kinase; Hydroxymethylglutaryl-CoA reductase kinase; prkaa2; protein kinase, AMP-activated, alpha 1 catalytic subunit; protein kinase, AMP-activated, alpha 2 catalytic subunit; Tau-protein kinase PRKAA1
Gene Aliases: 2310008I11Rik; A830082D05; AI194361; AI450832; AL024255; AMPK; AMPK1; AMPK2; AMPKa1; AMPKa2; AMPKalpha1; AMPKalpha2; C130083N04Rik; PRKAA; PRKAA1; PRKAA2
UniProt ID: (Human) Q13131, (Human) P54646, (Rat) P54645, (Mouse) Q5EG47, (Rat) Q09137, (Mouse) Q8BRK8
Entrez Gene ID: (Human) 5562, (Human) 5563, (Rat) 65248, (Mouse) 105787, (Rat) 78975, (Mouse) 108079
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.