Invitrogen
Manufacturer Code:PA595669
Catalog # PIPA595669
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Fruit fly |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic phospho-peptide corresponding to amino acid residues surrounding Thr342 conjugated to KLH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Akt; Akt kinase; Akt1-PA; Akt1-PB; Akt1-PC; Akt1-PD; Akt1-PE; CG4006-PA; CG4006-PB; CG4006-PC; CG4006-PD; CG4006-PE; DAkt; dAkt kinase; PKB; Protein kinase B; RAC serine/threonine protein kinase; RAC serine/threonine-protein kinase; related to PKA to PKC protein kinases; related to the A and C kinases; SD10374p
Gene Aliases: AKT; AKT/PKB; AKT1; Akt1|PKB; CG4006; D-Akt; DAKT; dAKT/dPKB; dAkt/PKB; DAKT1; DAKT1/PKB; Dmel\CG4006; Dmel_CG4006; DPKB; DRAC-PK; DRAC-PK66; DRAC-PK85; l(3)04226; l(3)89Bq; P-AKT; pAkt; PKB; PKB/AKT; PKB/dAKT; RAC; RacPK
UniProt ID: (Fruit fly) Q8INB9
Entrez Gene ID: (Fruit fly) 41957
Molecular Function: annexin calcium-binding protein calmodulin intracellular calcium-sensing protein kinase non-receptor serine/threonine protein kinase protein kinase transfer/carrier protein transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.