Invitrogen
Manufacturer Code:PA536821
Catalog # PIPA536821
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic phosphopeptide derived from human ABL1/2 around the phosphorylation site of Tyrosine 393/439 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Abelson murine leukemia oncogene; Abelson murine leukemia viral (v-abl) oncogene homolog 1; Abelson murine leukemia viral (v-abl) oncogene homolog 2; Abelson murine leukemia viral oncogene homolog 1; Abelson murine leukemia viral oncogene homolog 2; Abelson tyrosine-protein kinase 1; Abelson tyrosine-protein kinase 2; Abelson-related gene protein; ABL1 ABL2 ABL JTK7 p150 Proto-oncogene c-Abl Tyrosine-protein kinase ABL1 c-Abl Abelson tyrosine-protein kinase 2 Abelson murine leukemia viral oncogene homolog 2 Abelson-related gene protein Tyrosine-protein kinase ARG ABLL ARG; Abl1-related gene product; abl2; arg tyrosine kinase; bcr/c-abl oncogene protein; c-abl oncogene 1, non-receptor tyrosine kinase; c-abl oncogene 1, receptor tyrosine kinase; c-abl oncogene 2, non-receptor tyrosine kinase; p150; Proto-oncogene c-Abl; proto-oncogene tyrosine-protein kinase ABL1; tyrosine kinase ARG; Tyrosine-protein kinase ABL1; Tyrosine-protein kinase ABL2; Tyrosine-protein kinase ARG; v-abl Abelson murine leukemia oncogene 1; v-abl Abelson murine leukemia viral oncogene 1; v-abl Abelson murine leukemia viral oncogene 2; v-abl Abelson murine leukemia viral oncogene homolog 1; v-abl Abelson murine leukemia viral oncogene homolog 2; v-abl Abelson murine leukemia viral oncogene homolog 2 (arg, Abelson-related gene)
Gene Aliases: AA536808; ABL; ABL1; ABL2; ABLL; AI325092; ARG; bcr/abl; c-ABL; c-ABL1; E430008G22Rik; JTK7; p150; v-abl
UniProt ID: (Human) P00519, (Human) P42684, (Mouse) P00520
Entrez Gene ID: (Human) 25, (Human) 27, (Rat) 311860, (Mouse) 11350, (Mouse) 11352, (Rat) 304883
Molecular Function:
kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.