Invitrogen
Manufacturer Code:PA537764
Catalog # PIPA537764
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide sequence around phosphorylation site of serine 58 (R-S-S(p)-W-R) derived from Human 14-3-3 zeta. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 14-3-3 delta; 14-3-3 protein zeta/delta; 14-3-3 protein zeta/delta 1433Z 143Z FAS Factor activating exoenzyme S KCIP-1 Mitochondrial import stimulation factor S1 subunit Protein kinase C inhibitor protein-1 YWHAZ; 14-3-3 protein/cytosolic phospholipase A2; 14-3-3 zeta; epididymis luminal protein 4; epididymis secretory protein Li 3; epididymis secretory protein Li 93; KCIP-1; Mitochondrial import stimulation factor S1 subunit; phospholipase A2; Protein kinase C inhibitor protein 1; protein kinase C inhibitor protein-1; SEZ-2; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, delta polypeptide; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, zeta polypeptide; tyrosine 3/tryptophan 5 -monooxygenase activation protein, zeta polypeptide
Gene Aliases: 1110013I11Rik; 14-3-3-zeta; 14-3-3z; 14-3-3zeta; AI596267; AL022924; AU020854; HEL-S-3; HEL-S-93; HEL4; KCIP-1; Msfs1; YWHAD; YWHAZ
UniProt ID: (Human) P63104, (Mouse) P63101, (Rat) P63102
Entrez Gene ID: (Human) 7534, (Mouse) 22631, (Rat) 25578
Molecular Function: chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.