Novus Biologicals
Manufacturer Code:NBP293747100UL
Catalog # NB029356
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-246 of human PDC (NP_002588.3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 33 kDa phototransducing protein; 33 kDa phototransducing protein 33kDA phototransducing protein G beta gamma binding protein PHD PhLP phosducin phosducin-like orphan protein; G beta gamma binding protein; PHD; Phosducin; phosducin-like orphan protein; Protein MEKA
Gene Aliases: MEKA; PDC; PHD; PhLOP; PhLP
UniProt ID: (Human) P20941
Entrez Gene ID: (Human) 5132
Molecular Function:
G-protein modulator
chaperone
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.