Abnova
Manufacturer Code:PAB5919
Catalog # 89-132-006
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to C-terminus of mouse Pfkfb3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Pfkfb3.
For Research Use Only
Protein Aliases: 6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase 3; E330010H22Rik; inducible 6-phosphofructo-2-kinase; iPFK-2; uPFK-2
Gene Aliases: E330010H22Rik; iPFK-2; uPFK-2
Entrez Gene ID: (Mouse) 170768
Molecular Function:
carbohydrate phosphatase
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.