Novus Biologicals
Manufacturer Code:NB100242
Catalog # NB100242
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 475 and 525 of mouse Pescadillo using the numbering given in entry NP_075027.1 (GeneID 64934). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PES1; Pescadillo homolog; pescadillo homolog 1, containing BRCT domain; pescadillo homolog pescadillo homolog 1 containing BRCT domain (zebrafish) PESpescadillo (zebrafish) homolog 1 containing BRCT domain
Gene Aliases: PES; PES1
UniProt ID: (Human) O00541
Entrez Gene ID: (Human) 23481
Molecular Function: RNA binding protein nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.