Novus Biologicals
Manufacturer Code:NBP299498AF488
Catalog # NB098886
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human Peroxiredoxin 4. |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Antioxidant enzyme AOE372; Antioxidant enzyme AOE372 AOE37-2PRX-4 EC 1.11.1.15 peroxiredoxin 4 Peroxiredoxin IV peroxiredoxin-4 prx4 prx-4 Prx-IV thioredoxin peroxidase (antioxidant enzyme) Thioredoxin peroxidase AO372 Thioredoxin-dependent peroxide reductase A0372; AOE37-2; epididymis secretory sperm binding protein Li 97n; Peroxiredoxin IV; Peroxiredoxin-4; Prx-IV; thioredoxin peroxidase (antioxidant enzyme); Thioredoxin peroxidase AO372; Thioredoxin-dependent peroxide reductase A0372; Thioredoxin-dependent peroxiredoxin 4
Gene Aliases: AOE37-2; AOE372; HEL-S-97n; PRDX4; PRX-4
UniProt ID: (Human) Q13162
Entrez Gene ID: (Human) 10549
Molecular Function: oxidoreductase peroxidase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.