R&D Systems
Manufacturer Code:MAB3489SP
Catalog # MAB3489SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 477719 |
Immunogen | <EM>E. coli</EM>-derived recombinant human Peroxiredoxin 2 Met1-Asn198 Accession # P32119 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 1.11.1 MGC4104 Natural killer cell-enhancing factor B natural killer-enhancing factor B NKEF-B NKEFBNKEF-B peroxiredoxin 2 peroxiredoxin-2 PRDX2 PRPTDPX1 PRX2 PRXII TDPX1 thiol-specific antioxidant 1 Thiol-specific antioxidant protein Thioredoxin peroxidase 1 Thioredoxin-dependent peroxide reductase 1 torin TPX1 TSA TSAEC 1.11.1.15; epididymis secretory sperm binding protein Li 2a; Natural killer cell-enhancing factor B; NKEF-B; Peroxiredoxin-2; PRP; thiol-specific antioxidant 1; Thiol-specific antioxidant protein; Thioredoxin peroxidase 1; Thioredoxin-dependent peroxide reductase 1; Thioredoxin-dependent peroxiredoxin 2; torin; TSA
Gene Aliases: HEL-S-2a; NKEF-B; NKEFB; PRDX2; PRP; PRX2; PRXII; PTX1; TDPX1; TPX1; TSA
UniProt ID: (Human) P32119
Entrez Gene ID: (Human) 7001
Molecular Function:
oxidoreductase
peroxidase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.