Novus Biologicals
Manufacturer Code:NBP154337
Catalog # NBP154337
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to PRDX2(peroxiredoxin 2) The peptide sequence was selected from the middle region of PRDX2. Peptide sequence VLGVSVDSQFTHLAWINTPRKEGGLGPLNIPLLADVTRRLSEDYGVLKTD. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 1.11.1 MGC4104 Natural killer cell-enhancing factor B natural killer-enhancing factor B NKEFBNKEF-B peroxiredoxin 2 peroxiredoxin-2 PRPTDPX1 PRX2 PRXII thiol-specific antioxidant 1 Thiol-specific antioxidant protein Thioredoxin peroxidase 1 Thioredoxin-dependent peroxide reductase 1 torin TPX1 TSAEC 1.11.1.15; epididymis secretory sperm binding protein Li 2a; Natural killer cell-enhancing factor B; NKEF-B; Peroxiredoxin-2; PRP; thiol-specific antioxidant 1; Thiol-specific antioxidant protein; Thioredoxin peroxidase 1; Thioredoxin-dependent peroxide reductase 1; Thioredoxin-dependent peroxiredoxin 2; torin; TSA
Gene Aliases: HEL-S-2a; NKEF-B; NKEFB; PRDX2; PRP; PRX2; PRXII; PTX1; TDPX1; TPX1; TSA
UniProt ID: (Human) P32119
Entrez Gene ID: (Human) 7001
Molecular Function:
oxidoreductase
peroxidase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.