Proteintech
Manufacturer Code:198991AP150UL
Catalog # 50-173-2590
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Periostin Fusion Protein Ag13791 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: OSF 2 OSF2 Osteoblast specific factor 2 PDLPOSTN Periostin PN POSTN RP11 412K4.1; OSF-2; osteoblast specific factor 2 (fasciclin I-like); Osteoblast-specific factor 2; periodontal ligament-specific periostin; Periostin; periostin, osteoblast specific factor; periostin-like factor; PN
Gene Aliases: A630052E07Rik; AI747096; OSF-2; OSF2; PDLPOSTN; peri; PLF; PN; POSTN
UniProt ID: (Human) Q15063, (Mouse) Q62009
Entrez Gene ID: (Human) 10631, (Rat) 361945, (Mouse) 50706
Molecular Function:
cell adhesion molecule
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.