Proteintech
Manufacturer Code:269511AP150UL
Catalog # 50-173-5514
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Perilipin 5 Fusion Protein Ag25272 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: lipid droplet associated protein; Lipid droplet-associated protein PAT-1; Lipid storage droplet protein 5; LSDP5 OXPAT Perilipin 5 PLIN5; Myocardial LD protein; Perilipin-5
Gene Aliases: 2310076L09Rik; AI415325; AW109675; LSDA5; LSDP5; MLDP; OXPAT; PAT-1; Pat1; PLIN5
UniProt ID: (Human) Q00G26, (Mouse) Q8BVZ1
Entrez Gene ID: (Human) 440503, (Pig) 100142670, (Mouse) 66968
Molecular Function:
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.