Invitrogen
Manufacturer Code:12939280
Catalog # 50-112-63
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | eBioOMAK-D |
Conjugate | PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: Cytolysin; cytolysin FLH2 HPLH2 lymphocyte pore forming protein lymphocyte pore-forming protein perforin 1 (pore forming protein) perforin-1 PFN1 PFP RP11-710A11.3; Lymphocyte pore-forming protein; P1; Perforin-1; pore forming protein
Gene Aliases: Pfn; Pfp; Prf-1; Prf1
UniProt ID: (Mouse) P10820
Entrez Gene ID: (Mouse) 18646
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.