Invitrogen
Manufacturer Code:12999442
Catalog # 50-113-54
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | dG9 (delta G9) |
Conjugate | PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Cytolysin; cytolysin FLH2 HPLH2 lymphocyte pore forming protein lymphocyte pore-forming protein perforin 1 (pore forming protein) perforin-1 PFN1 PFP RP11-710A11.3; lymphocyte pore forming protein; Lymphocyte pore-forming protein; P1; perforin 1 (pore forming protein); Perforin-1; PFP; pore-forming protein
Gene Aliases: FLH2; HPLH2; P1; PFN1; PFP; PRF1
UniProt ID: (Human) P14222
Entrez Gene ID: (Human) 5551, (Pig) 396595
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.