R&D Systems
Manufacturer Code:MAB8011100
Catalog # MAB8011100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1001103 |
Immunogen | Synthetic peptide containing human Perforin Pro22-Trp555 Accession No. P14222 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 °C as supplied. 1 month 2 to 8 °C under sterile conditions after reconstitution. 6 months -20 to -70 °C under sterile condition |
Mouse Monoclonal Antibody
Protein Aliases: Cytolysin; Cytolysin FLH2 HPLH2 HPLH2lymphocyte pore forming protein Lymphocyte pore-forming protein MGC65093 P1 P1PFN1 perforin 1 (pore forming protein) perforin-1 PFP PFPcytolysin PRF1; lymphocyte pore forming protein; Lymphocyte pore-forming protein; P1; perforin 1 (pore forming protein); Perforin-1; PFP
Gene Aliases: FLH2; HPLH2; P1; PFN1; PFP; PRF1
UniProt ID: (Human) P14222
Entrez Gene ID: (Human) 5551
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.