Invitrogen
Manufacturer Code:PA579865
Catalog # PIPA579865
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E.coli-derived human Perforin recombit protein (Position: E175-W555). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cytolysin; lymphocyte pore forming protein; Lymphocyte pore-forming protein; P1; perforin 1 (pore forming protein); Perforin-1; Perforin-1 P1 Cytolysin Lymphocyte pore-forming protein PFP PRF1 PFP; PFP
Gene Aliases: FLH2; HPLH2; P1; PFN1; PFP; PRF1
UniProt ID: (Human) P14222
Entrez Gene ID: (Human) 5551
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.