Invitrogen
Manufacturer Code:PA517431
Catalog # PIPA517431
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues surrounding leucine 349 of mouse perforin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cytolysin; Lymphocyte pore-forming protein; OTTHUMP00000019759 cytolysin lymphocyte pore forming protein lymphocyte pore-forming protein perforin-1; P1; Perforin-1; pore forming protein
Gene Aliases: Pfn; Pfp; Prf-1; Prf1
UniProt ID: (Mouse) P10820
Entrez Gene ID: (Mouse) 18646
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.