R&D Systems
Manufacturer Code:MAB21661
Catalog # MAB21661
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 265641 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse Pentraxin 3 Glu18-Ser381 Accession # P48759 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: alpha-induced protein 5 pentaxin-related gene rapidly induced by IL-1 beta tumor necrosis factor Pentaxin-related protein PTX3 pentraxin 3 long pentraxin-3 pentraxin-related gene rapidly induced by IL-1 beta pentraxin-related protein PTX3 PTX3 TNF alpha-induced protein 5 TNFAIP5 TSG14 TSG-14 TSG-14pentaxin-related gene rapidly induced by IL-1 beta Tumor necrosis factor alpha-induced protein 5 tumor necrosis factor alpha-induced protein 5 Tumor necrosis factor-inducible gene 14 protein tumor necrosis factor-inducible protein TSG-14; long pentraxin 3; Pentaxin-related protein PTX3; Pentraxin-related protein PTX3; TNF alpha-induced protein 5; TSG-14; Tumor necrosis factor alpha-induced protein 5; Tumor necrosis factor-inducible gene 14 protein; tumor necrosis factor-inducible protein TSG-14
Gene Aliases: PTX3; TNFAIP5; TSG-14; TSG14
UniProt ID: (Human) P26022
Entrez Gene ID: (Human) 5806
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.