Proteintech
Manufacturer Code:137971AP150UL
Catalog # 50-172-9013
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Pentraxin 3 Fusion Protein Ag4624 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: long pentraxin 3; pentaxin related; Pentaxin related protein PTX3 Pentraxin 3 Pentraxin related protein PTX3 PTX3 TNF alpha induced protein 5 TNFAIP5 TSG 14 TSG14; Pentaxin-related protein PTX3; pentraxin 3; pentraxin related; Pentraxin-related protein PTX3; TNF alpha-induced protein 5; TSG-14; Tumor necrosis factor alpha-induced protein 5; Tumor necrosis factor-inducible gene 14 protein; tumor necrosis factor-inducible protein TSG-14
Gene Aliases: AI607804; PTX3; TNFAIP5; TSG-14; TSG14
UniProt ID: (Human) P26022, (Mouse) P48759
Entrez Gene ID: (Human) 5806, (Mouse) 19288, (Rat) 689388
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.