Novus Biologicals
Manufacturer Code:NBP273334F
Catalog # NBP273334F
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1D6 |
Immunogen | Full length recombinant protein of human Pentraxin 2/SAP (NP_001630) produced in HEK293T cells. |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: 9.5S alpha-1-glycoprotein; 9.5S alpha-1-glycoprotein amyloid P component serum MGC88159 PTX2serum amyloid P-component SAPpentaxin-related Serum Amyloid P component; epididymis secretory sperm binding protein Li 92n; pentaxin-related; pentraxin-2; pentraxin-related; SAP; Serum amyloid P-component; Serum amyloid P-component(1-203)
Gene Aliases: APCS; HEL-S-92n; PTX2; SAP
UniProt ID: (Human) P02743
Entrez Gene ID: (Human) 325
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.