Abnova
Manufacturer Code:PAB11572
Catalog # 89-115-188
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to internal region of mouse Penk. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of Penk.
For Research Use Only
Protein Aliases: AI326464; ENK; Leu-enkephalin; Met-enkephalin; Met-enkephalin-Arg-Phe; Met-enkephalin-Arg-Ser-Leu; OGF; Opioid growth factor; Penk; PENK(114-133); PENK(143-184); PENK(238-259); PPA; preproenkephalin 1; Proenkephalin-A; spermatogenic-specific proenkephalin; Synenkephalin
Gene Aliases: AI326464; ENK; Penk; Penk1; PPA
UniProt ID: (Mouse) P22005
Entrez Gene ID: (Mouse) 18619
Molecular Function: neuropeptide peptide hormone signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.