Invitrogen
Manufacturer Code:PA558573
Catalog # PIPA558573
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human Pellino 3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E3 ubiquitin-protein ligase pellino homolog 3; E3 ubiquitin-protein ligase pellino homolog 3 Pellino-3 632- PELI3; pellino homolog 3; Pellino-3; protein pellino homolog 3; RING-type E3 ubiquitin transferase pellino homolog 3
Gene Aliases: PELI3
UniProt ID: (Human) Q8N2H9
Entrez Gene ID: (Human) 246330
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.