Invitrogen
Manufacturer Code:PA544181
Catalog # PIPA544181
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human PELI3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E3 ubiquitin-protein ligase pellino homolog 3; E3 ubiquitin-protein ligase pellino homolog 3 pseudogene; pellino 3; Pellino E3 ubiquitin protein ligase family member 3 MGC35521; pellino homolog 3; Pellino-3; protein pellino homolog 3; RING-type E3 ubiquitin transferase pellino homolog 3
Gene Aliases: 6030441F14Rik; A930011L17; BC028931; PELI3; RGD1305989
UniProt ID: (Human) Q8N2H9, (Mouse) Q8BXR6
Entrez Gene ID: (Rabbit) 100352313, (Horse) 100052587, (Bovine) 504839, (Human) 246330, (Pig) 100519073, (Rat) 309157, (Mouse) 240518, (Guinea pig) 100716351
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.