Invitrogen
Manufacturer Code:PA534465
Catalog # PIPA534465
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 15 amino acid peptide near the carboxy terminus of human Pellino 1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E3 ubiquitin-protein ligase pellino homolog 1; pellino 1; Pellino 1 Pellino E3 ubiquitin protein ligase 1 PELI1 pellino1 pellino-1; pellino homolog 1; Pellino-1; Pellino-related intracellular-signaling molecule; protein pellino homolog 1; RING-type E3 ubiquitin transferase pellino homolog 1
Gene Aliases: 2810468L03Rik; A930031K15Rik; AA409794; AI586297; D11Ertd676e; PELI1; PRISM
UniProt ID: (Human) Q96FA3, (Mouse) Q8C669
Entrez Gene ID: (Human) 57162, (Mouse) 67245, (Rat) 305549
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.