Abnova
Manufacturer Code:PAB17984
Catalog # 89-119-592
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to mouse Pdgfb. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant Pdgfb.
For Research Use Only
Protein Aliases: c-sis; PDGF subunit B; PDGF-2; PDGF-B; Platelet-derived growth factor B chain; Platelet-derived growth factor beta polypeptide; Platelet-derived growth factor subunit B; Proto-oncogene c-Sis; Sis
Gene Aliases: PDGF-B; Pdgfb; Sis
UniProt ID: (Mouse) P31240
Entrez Gene ID: (Mouse) 18591
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.