Abnova
Manufacturer Code:PAB24719
Catalog # 89-126-384
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to mouse Pde1a. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Pde1a.
For Research Use Only
Protein Aliases: 61 kDa Cam-PDE; AI987702; AW125737; Calcium/calmodulin-dependent 3',5'-cyclic nucleotide phosphodiesterase 1A; Cam-PDE 1A; MGC116577
Gene Aliases: AI987702; AW125737; Pde1a
UniProt ID: (Mouse) Q61481
Entrez Gene ID: (Mouse) 18573
Molecular Function:
hydrolase
phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.