Novus Biologicals
Manufacturer Code:NBP281063DL594
Catalog # NB015194
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | LD4 sAB |
Immunogen | Peptide corresponding to human paxillin-LD4 motif (residues 260-278) fused to MBP peptide sequence: WGGSATRELDELMASLSD. |
Conjugate | DyLight 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: FLJ16691 paxillin; Paxillin; testicular tissue protein Li 134
Gene Aliases: PXN
UniProt ID: (Human) P49023
Entrez Gene ID: (Human) 5829
Molecular Function: actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.