Invitrogen
Manufacturer Code:MA535261
Catalog # PIMA535261
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0368 |
Immunogen | A synthesized peptide derived from human Paxillin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: AW108311 AW123232 FLJ16691 hCG_1778014 I79_001229 Pax PAX-1 PAXI Paxillin paxillin alpha PXN wu:fw71f12; myocardial ischemic preconditioning associated protein 7; Paxillin; testicular tissue protein Li 134
Gene Aliases: AW108311; AW123232; Pax; PXN
UniProt ID: (Human) P49023, (Mouse) Q8VI36, (Rat) Q66H76
Entrez Gene ID: (Human) 5829, (Mouse) 19303, (Rat) 360820
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.