R&D Systems
Manufacturer Code:MAB3487SP
Catalog # MAB3487SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 1207C |
Immunogen | <em>E. coli</em>-derived recombinant human Pax5 Thr141-His391 Accession # Q02548 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: B cell specific activator protein B-cell-specific transcription factor BSAP BSAPB-cell lineage specific activator paired box 5 paired box gene 5 (B-cell lineage specific activator protein) paired box gene 5 (B-cell lineage specific activator) paired box homeotic gene 5 paired box protein Pax-5 transcription factor PAX 5; B-cell lineage specific activator; B-cell-specific transcription factor; BSAP; paired box homeotic gene 5; Paired box protein Pax-5; paired domain gene 5; transcription factor PAX 5
Gene Aliases: ALL3; BSAP; PAX5
UniProt ID: (Human) Q02548
Entrez Gene ID: (Human) 5079
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.