Invitrogen
Manufacturer Code:12991880
Catalog # 50-112-87
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 1H9 |
Conjugate | PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: B-cell lineage specific activator; B-cell-specific transcription factor; BSAP; paired box gene 5; paired box homeotic gene 5; Paired box protein Pax-5; paired domain gene 5; Pax5 variant 1; Pax5 variant 2; transcription factor PAX 5
Gene Aliases: ALL3; BSAP; EBB-1; KLP; Pax-5; PAX5
UniProt ID: (Human) Q02548, (Mouse) Q02650
Entrez Gene ID: (Human) 5079, (Mouse) 18507
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.