OriGene
Manufacturer Code:TA803822
Catalog # 50-167-9942
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5G9 |
Immunogen | Human recombit protein fragment corresponding to amino acids 770-1027 of human PTCH1 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: BCNS NBCCS PTC PTC1 PTCH; Protein patched homolog 1; PTC; PTCH protein +12b; PTCH protein +4'; PTCH protein -10; PTCH protein -3,4,5
Gene Aliases: BCNS; HPE7; NBCCS; PTC; PTC1; PTCH; PTCH1; PTCH11
UniProt ID: (Human) Q13635
Entrez Gene ID: (Human) 5727
Molecular Function:
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.