R&D Systems
Manufacturer Code:MAB4078
Catalog # MAB4078
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 403438 |
Immunogen | <EM>E. coli</EM>-derived recombinant human Patched 2/PTCH2 Glu79-Glu392 Accession # Q9Y6C5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: patched (Drosophila) homolog 2 patched 2 patched homolog 2 patched homolog 2 (Drosophila) protein patched homolog 2 ptc2 PTCH2; patched homolog 2; Protein patched homolog 2; PTC2
Gene Aliases: PTC2; PTCH2; UNQ560/PRO1121/PRO57079
UniProt ID: (Human) Q9Y6C5
Entrez Gene ID: (Human) 8643
Molecular Function:
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.