Abnova
Manufacturer Code:PAB9559
Catalog # 89-134-230
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to C-terminus residues surrounding Asp214 of mouse Parp1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Parp1.
For Research Use Only
Protein Aliases: 5830444G22Rik; ADP-ribosyltransferase (NAD+, poly (ADP-ribose) polymerase) 1; ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase) 1; ADP-ribosyltransferase diphtheria toxin-like 1; Adprp; ADPRT 1; Adprt1; AI893648; C80510; msPARP; NAD(+) ADP-ribosyltransferase 1; PARP; parp-1; poly [ADP-ribose] polymerase 1; poly[ADP-ribose] synthase 1; poly[ADP-ribose] synthetase 1; PPOL; sPARP-1
Gene Aliases: 5830444G22Rik; Adprp; Adprt1; AI893648; ARTD1; C80510; PARP; parp-1; PPOL; sPARP-1
Entrez Gene ID: (Mouse) 11545
Molecular Function: DNA binding protein DNA ligase ligase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.