Novus Biologicals
Manufacturer Code:NBP2298380.025ML
Catalog # NB019485
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | PRK8 |
Immunogen | Full length recombinant human Parkin. Epitope has been mapped to the second ring domain (aa 399-465). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: E3 ubiquitin ligase E3 ubiquitin-protein ligase parkin EC 6.3.2.- LPRS2 parkin parkin 2 Parkinson disease protein 2 Parkinson juvenile disease protein 2 parkinson protein 2 E3 ubiquitin protein ligase (parkin) PDJjuvenile) 2 parkin; E3 ubiquitin-protein ligase parkin; Parkin; Parkin RBR E3 ubiquitin-protein ligase; Parkinson disease (autosomal recessive, juvenile) 2, parkin; Parkinson disease protein 2; Parkinson juvenile disease protein 2; parkinson protein 2 E3 ubiquitin protein ligase; parkinson protein 2, E3 ubiquitin protein ligase (parkin)
Gene Aliases: AR-JP; LPRS2; PARK2; PDJ; PRKN
UniProt ID: (Human) O60260
Entrez Gene ID: (Human) 5071
Molecular Function: ligase ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.