Invitrogen
Manufacturer Code:PA513420
Catalog # PIPA513420
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Fruit fly |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 82-111 amino acids from the N-terminal region of Drosophila Parkin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CG10523-PB; CG10523-PB isoform B Cg10523-pc isoform c PARKIN park parkin; CG10523-PC; D-parkin; dparkin; E3 ubiquitin-protein ligase parkin; park-PB; park-PC; parkin F
Gene Aliases: AAN12155.1; CG10523; Dmel\CG10523; Dmel_CG10523; Dpark; dPARKIN; dpk; PARK; park1; PARK2; PARKIN; PK; SD01679
UniProt ID: (Fruit fly) Q7KTX7
Entrez Gene ID: (Fruit fly) 40336
Molecular Function:
ligase
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.