R&D Systems
Manufacturer Code:AF1438SP
Catalog # AF1438SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human Parkin-2 Met1-Val437 Accession # NP_054642 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: AR-JP E3 ubiquitin ligase E3 ubiquitin-protein ligase parkin EC 6.3.2.- LPRS2 PARK2 parkin parkin 2 Parkinson disease protein 2 Parkinson juvenile disease protein 2 parkinson protein 2 E3 ubiquitin protein ligase (parkin) PDJ PDJjuvenile) 2 parkin PRKN; E3 ubiquitin-protein ligase parkin; Parkin; Parkin RBR E3 ubiquitin-protein ligase; Parkinson disease (autosomal recessive, juvenile) 2, parkin; Parkinson disease protein 2; Parkinson juvenile disease protein 2; parkinson protein 2 E3 ubiquitin protein ligase; parkinson protein 2, E3 ubiquitin protein ligase (parkin)
Gene Aliases: AR-JP; LPRS2; PARK2; PDJ; PRKN
UniProt ID: (Human) O60260
Entrez Gene ID: (Human) 5071
Molecular Function: ligase ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.